α,α,6-Trimethyl-1,3-benzodioxole-5-methanol ≥95%

Supplier: AOBChem USA

Warning

191722-1G 191722-500MG 191722-5G
77466-484EA 600.49 USD
77466-484 77479-228 77489-464
α,α,6-Trimethyl-1,3-benzodioxole-5-methanol ≥95%
α,α,6-Trimethyl-1,3-benzodioxole-5-methanol

Precaución: For research use only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34861852
CAS Number: 1780228-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR