2-[(4-Bromo-2-fluorophenyl)methyl]oxirane ≥95%

Supplier: AOBChem USA

Warning

190411-1G 190411-500MG 190411-5G 190411-25G
77466-476EA 423.33 USD
77466-476 77479-220 77489-458 77499-806
2-[(4-Bromo-2-fluorophenyl)methyl]oxirane ≥95%
2-[(4-Bromo-2-fluorophenyl)methyl]oxirane

Precaución: For research use only

Formula: C₉H₈BrFO
MW: 231.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34861172
CAS Number: 2228313-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR