2-[(2-Fluorophenyl)methyl]oxirane ≥95%

Supplier: AOBChem USA

Warning

190329-1G 190329-500MG 190329-5G 190329-25G
77466-472EA 376.98 USD
77466-472 77479-216 77489-454 77499-802
2-[(2-Fluorophenyl)methyl]oxirane ≥95%
2-[(2-Fluorophenyl)methyl]oxirane

Precaución: For research use only

Formula: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Ambient
CAS Number: 67959-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR