2-[(4-Bromophenyl)methyl]oxirane ≥96%

Supplier: AOBChem USA

Warning

190304-1G 190304-500MG 190304-5G
77466-468EA 774.56 USD
77466-468 77479-212 77489-450
2-[(4-Bromophenyl)methyl]oxirane ≥96%
2-[(4-Bromophenyl)methyl]oxirane

Precaución: For research use only

Formula: C₉H₉BrO
MW: 213.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31431895
CAS Number: 62826-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR