α-(Difluoromethyl)benzenemethanol ≥96%

Supplier: AOBChem USA

Warning

189735-1G 189735-500MG 189735-5G 189735-25G
77466-464EA 168.92 USD
77466-464 77479-208 77489-446 77499-798
α-(Difluoromethyl)benzenemethanol ≥96%
α-(Difluoromethyl)benzenemethanol

Precaución: For research use only

Formula: C₈H₈F₂O
MW: 158.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779818
CAS Number: 345-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR