5-Bromo-2-fluoro-1-(methylthio)-3-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189363-1G 189363-500MG 189363-5G 189363-25G
77466-456EA 415.09 USD
77466-456 77479-200 77489-438 77499-794
5-Bromo-2-fluoro-1-(methylthio)-3-(triethylsilyl)benzene ≥95%
5-Bromo-2-fluoro-1-(methylthio)-3-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₃H₂₀BrFSSi
Storage Temperature: Ambient
MDL Number: MFCD34860725
CAS Number: 2266623-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR