1,3-Difluoro-4-iodo-2-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189356-1G 189356-500MG 189356-5G 189356-25G
77466-452EA 415.09 USD
77466-452 77479-196 77489-434 77499-790
1,3-Difluoro-4-iodo-2-(triethylsilyl)benzene ≥95%
1,3-Difluoro-4-iodo-2-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₂H₁₇F₂ISi
MW: 354.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30179092
CAS Number: 2185517-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR