1-(Methylthio)-3-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189334-1G 189334-500MG 189334-5G 189334-25G
77466-448EA 415.09 USD
77466-448 77479-192 77489-430 77499-786
1-(Methylthio)-3-(triethylsilyl)benzene ≥95%
1-(Methylthio)-3-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₃H₂₂SSi
MW: 238.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860705
CAS Number: 2842852-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR