1-(1,1-Dimethylethyl)-3-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189312-1G 189312-500MG 189312-5G 189312-25G
77466-444EA 415.09 USD
77466-444 77479-188 77489-426 77499-782
1-(1,1-Dimethylethyl)-3-(triethylsilyl)benzene ≥95%
1-(1,1-Dimethylethyl)-3-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₆H₂₈Si
MW: 248.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860686
CAS Number: 17897-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR