1-Fluoro-3-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189302-1G 189302-500MG 189302-5G 189302-25G
77466-440EA 415.09 USD
77466-440 77479-184 77489-422 77499-778
1-Fluoro-3-(triethylsilyl)benzene ≥95%
1-Fluoro-3-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₂H₁₉FSi
MW: 210.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860678
CAS Number: 1868070-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR