2,4-Dimethyl-1-(triethylsilyl)benzene ≥97%

Supplier: AOBChem USA

Warning

189296-1G 189296-500MG 189296-5G 189296-25G
77466-436EA 415.09 USD
77466-436 77479-180 77489-418 77499-774
2,4-Dimethyl-1-(triethylsilyl)benzene ≥97%
2,4-Dimethyl-1-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₄H₂₄Si
MW: 220.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860673
CAS Number: 870484-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR