1-(Triethylsilyl)-3-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189293-1G 189293-500MG 189293-5G 189293-25G
77466-432EA 415.09 USD
77466-432 77479-176 77489-414 77499-770
1-(Triethylsilyl)-3-(trifluoromethyl)benzene ≥95%
1-(Triethylsilyl)-3-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₃H₁₉F₃Si
MW: 260.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860670
CAS Number: 204391-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR