1-Fluoro-4-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189287-1G 189287-500MG 189287-5G 189287-25G
77466-428EA 415.09 USD
77466-428 77479-172 77489-410 77499-766
1-Fluoro-4-(triethylsilyl)benzene ≥95%
1-Fluoro-4-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₂H₁₉FSi
MW: 210.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860665
CAS Number: 2129647-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR