2-(Triethylsilyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

189282-1G 189282-500MG 189282-5G 189282-25G
77466-424EA 415.09 USD
77466-424 77479-168 77489-406 77499-762
2-(Triethylsilyl)-1,1'-biphenyl ≥95%
2-(Triethylsilyl)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₈H₂₄Si
MW: 268.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094132
CAS Number: 195525-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR