1-(1,1-Dimethylethyl)-4-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189279-1G 189279-500MG 189279-5G 189279-25G
77466-420EA 415.09 USD
77466-420 77479-164 77489-402 77499-758
1-(1,1-Dimethylethyl)-4-(triethylsilyl)benzene ≥95%
1-(1,1-Dimethylethyl)-4-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₆H₂₈Si
MW: 248.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860659
CAS Number: 1554400-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR