4-(Triethylsilyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

189276-1G 189276-500MG 189276-5G 189276-25G
77466-416EA 415.09 USD
77466-416 77479-160 77489-398 77499-754
4-(Triethylsilyl)-1,1'-biphenyl ≥95%
4-(Triethylsilyl)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₈H₂₄Si
MW: 268.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860656
CAS Number: 18057-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR