1-Methyl-4-(triethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

189265-1G 189265-500MG 189265-5G 189265-25G
77466-412EA 415.09 USD
77466-412 77479-156 77489-394 77499-750
1-Methyl-4-(triethylsilyl)benzene ≥95%
1-Methyl-4-(triethylsilyl)benzene

Precaución: For research use only

Formula: C₁₃H₂₂Si
MW: 206.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00092290
CAS Number: 3644-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR