1,1-Dimethylethyl 3-(4-ethylphenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%

Supplier: AOBChem USA

Warning

189258-1G 189258-500MG 189258-5G 189258-25G
77466-408EA 415.09 USD
77466-408 77479-152 77489-390 77499-746
1,1-Dimethylethyl 3-(4-ethylphenyl)-3-hydroxy-1-azetidinecarboxylate ≥95%
1,1-Dimethylethyl 3-(4-ethylphenyl)-3-hydroxy-1-azetidinecarboxylate

Precaución: For research use only

Formula: C₁₆H₂₃NO₃
MW: 277.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860649
CAS Number: 1485130-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR