1,1-Dimethylethyl 3-hydroxy-3-(2-methoxyphenyl)-1-azetidinecarboxylate ≥96%

Supplier: AOBChem USA

Warning

189144-1G 189144-500MG 189144-5G 189144-25G
77466-404EA 410.97 USD
77466-404 77479-148 77489-386 77499-744
1,1-Dimethylethyl 3-hydroxy-3-(2-methoxyphenyl)-1-azetidinecarboxylate ≥96%
1,1-Dimethylethyl 3-hydroxy-3-(2-methoxyphenyl)-1-azetidinecarboxylate

Precaución: For research use only

Formula: C₁₅H₂₁NO₄
MW: 279.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30802706
CAS Number: 1496354-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR