1,1-Dimethylethyl 3-hydroxy-3-(2-methylphenyl)-1-azetidinecarboxylate ≥96%

Supplier: AOBChem USA

Warning

189133-1G 189133-500MG 189133-5G 189133-25G
77466-400EA 264.71 USD
77466-400 77479-144 77489-382 77499-740
1,1-Dimethylethyl 3-hydroxy-3-(2-methylphenyl)-1-azetidinecarboxylate ≥96%
1,1-Dimethylethyl 3-hydroxy-3-(2-methylphenyl)-1-azetidinecarboxylate

Precaución: For research use only

Formula: C₁₅H₂₁NO₃
MW: 263.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21081306
CAS Number: 1225439-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR