3-Methoxybenzophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (3-Methoxyphenyl)(phenyl)methanone

189090-1G 189090-500MG 189090-5G 189090-25G
77466-396EA 248.23 USD
77466-396 77479-140 77489-378 77499-736
3-Methoxybenzophenone ≥95%
3-Methoxybenzophenone

Precaución: For research use only

Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319936
CAS Number: 6136-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR