2-(4-Chlorophenyl)quinazolin-4(3H)-one ≥95%

Supplier: AOBChem USA

Warning

189025-1G 189025-500MG 189025-5G 189025-25G
77466-392EA 299.73 USD
77466-392 77479-136 77489-374 77499-732
2-(4-Chlorophenyl)quinazolin-4(3H)-one ≥95%
2-(4-Chlorophenyl)quinazolin-4(3H)-one

Precaución: For research use only

Formula: C₁₄H₉ClN₂O
MW: 256.6870 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01989876
CAS Number: 7455-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR