(4-(Benzyloxy)-3,5-dimethylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

188370-1G 188370-500MG 188370-5G 188370-25G
77466-388EA 415.09 USD
77466-388 77479-132 77489-368 77499-724
(4-(Benzyloxy)-3,5-dimethylphenyl)(methyl)sulfane ≥95%
(4-(Benzyloxy)-3,5-dimethylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₆H₁₈OS
MW: 258.379 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34860087
CAS Number: 2969426-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR