2,5-Dibromoisophthalic acid ≥97%

Supplier: AOBChem USA

Warning

187866-1G 187866-500MG 187866-5G 187866-25G
77466-384EA 123.6 USD
77466-384 77479-128 77489-364 77499-720
2,5-Dibromoisophthalic acid ≥97%
2,5-Dibromoisophthalic acid

Precaución: For research use only

Formula: C₈H₄Br₂O₄
MW: 323.923 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28719346
CAS Number: 379711-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR