tert-Butyl 3-(3-bromo-2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

187759-1G 187759-500MG 187759-5G 187759-25G
77466-380EA 369.77 USD
77466-380 77479-124 77489-360 77499-716
tert-Butyl 3-(3-bromo-2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate ≥95%
tert-Butyl 3-(3-bromo-2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate

Precaución: For research use only

Formula: C₁₄H₁₇BrFNO₃
MW: 346.1920 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34859584
CAS Number: 1695236-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR