tert-Butyl 3-(2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

187752-1G 187752-500MG 187752-5G
77466-376EA 563.41 USD
77466-376 77479-120 77489-356
tert-Butyl 3-(2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate ≥95%
tert-Butyl 3-(2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate

Precaución: For research use only

Formula: C₁₄H₁₈FNO₃
MW: 267.2960 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21596789
CAS Number: 1225439-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR