(3-Bromo-2,6-difluorophenyl)triethylsilane ≥97%

Supplier: AOBChem USA

Warning

187738-1G 187738-500MG 187738-5G 187738-25G
77466-372EA 415.09 USD
77466-372 77479-116 77489-352 77499-710
(3-Bromo-2,6-difluorophenyl)triethylsilane ≥97%
(3-Bromo-2,6-difluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₂H₁₇BrF₂Si
Storage Temperature: Ambient
MDL Number: MFCD27926971
CAS Number: 651027-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR