(2-Chloro-6-fluorophenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

187735-1G 187735-500MG 187735-5G 187735-25G
77466-368EA 415.09 USD
77466-368 77479-112 77489-348 77499-706
(2-Chloro-6-fluorophenyl)triethylsilane ≥95%
(2-Chloro-6-fluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₂H₁₈ClFSi
MW: 244.8082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34859582
CAS Number: 2640112-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR