Triethyl(phenyl)silane ≥97%

Supplier: AOBChem USA

Warning

187726-1G 187726-500MG 187726-5G 187726-25G
77466-364EA 415.09 USD
77466-364 77479-108 77489-344 77499-702
Triethyl(phenyl)silane ≥97%
Triethyl(phenyl)silane
Formula: C₁₂H₂₀Si
Storage Temperature: Ambient
MDL Number: MFCD34859576
CAS Number: 2987-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR