Methyl 2-fluoro-3-methoxy-5-Methyl-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

187674-1G 187674-500MG 187674-5G
77466-360EA 676.71 USD
77466-360 77479-104 77489-340
Methyl 2-fluoro-3-methoxy-5-Methyl-α-oxobenzeneacetate ≥95%
Methyl 2-fluoro-3-methoxy-5-Methyl-α-oxobenzeneacetate

Precaución: For research use only

Formula: C₁₁H₁₁FO₄
MW: 226.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34859547
CAS Number: 2122043-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR