4-(3-Fluorophenyl)-1-(phenylmethyl)-4-piperidinol ≥95%

Supplier: AOBChem USA

Warning

187077-1G 187077-500MG 187077-5G 187077-25G
77466-356EA 347.11 USD
77466-356 77479-100 77489-336 77499-698
4-(3-Fluorophenyl)-1-(phenylmethyl)-4-piperidinol ≥95%
4-(3-Fluorophenyl)-1-(phenylmethyl)-4-piperidinol

Precaución: For research use only

Formula: C₁₈H₂₀FNO
MW: 285.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689289
CAS Number: 75527-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR