α-Cyclohexyl-2,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186987-1G 186987-500MG 186987-5G 186987-25G
77466-352EA 415.09 USD
77466-352 77479-096 77489-332 77499-694
α-Cyclohexyl-2,5-dimethylbenzenemethanol ≥95%
α-Cyclohexyl-2,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD11937138
CAS Number: 1181646-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR