α-Cyclohexyl-3,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186983-1G 186983-500MG 186983-5G 186983-25G
77466-348EA 415.09 USD
77466-348 77479-092 77489-328 77499-690
α-Cyclohexyl-3,5-dimethylbenzenemethanol ≥95%
α-Cyclohexyl-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD12806388
CAS Number: 1183202-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR