α-Cyclohexyl-4-(2-methylpropyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186971-1G 186971-500MG 186971-5G 186971-25G
77466-344EA 415.09 USD
77466-344 77479-088 77489-324 77499-686
α-Cyclohexyl-4-(2-methylpropyl)benzenemethanol ≥95%
α-Cyclohexyl-4-(2-methylpropyl)benzenemethanol

Precaución: For research use only

Formula: C₁₇H₂₆O
Storage Temperature: Ambient
MDL Number: MFCD14531252
CAS Number: 1216294-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR