α-Cyclohexyl-2,4,6-trifluorobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

186954-1G 186954-500MG 186954-5G 186954-25G
77466-340EA 415.09 USD
77466-340 77479-084 77489-320 77499-682
α-Cyclohexyl-2,4,6-trifluorobenzenemethanol ≥97%
α-Cyclohexyl-2,4,6-trifluorobenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD16357603
CAS Number: 1285342-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR