α-Cyclohexyl-4-propoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186936-1G 186936-500MG 186936-5G 186936-25G
77466-336EA 415.09 USD
77466-336 77479-080 77489-316 77499-678
α-Cyclohexyl-4-propoxybenzenemethanol ≥95%
α-Cyclohexyl-4-propoxybenzenemethanol

Precaución: For research use only

Formula: C₁₆H₂₄O₂
Storage Temperature: Ambient
MDL Number: MFCD17126806
CAS Number: 1466307-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR