α-Cyclohexyl-4-methoxy-3,5-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

186913-1G 186913-500MG 186913-5G 186913-25G
77466-332EA 415.09 USD
77466-332 77479-076 77489-312 77499-674
α-Cyclohexyl-4-methoxy-3,5-dimethylbenzenemethanol ≥95%
α-Cyclohexyl-4-methoxy-3,5-dimethylbenzenemethanol

Precaución: For research use only

Formula: C₁₆H₂₄O₂
MW: 248.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25866169
CAS Number: 1558182-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR