α-Cyclohexyl-4-(ethylthio)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

186874-1G 186874-500MG 186874-5G 186874-25G
77466-328EA 415.09 USD
77466-328 77479-072 77489-308 77499-670
α-Cyclohexyl-4-(ethylthio)benzenemethanol ≥97%
α-Cyclohexyl-4-(ethylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₅H₂₂OS
Storage Temperature: Ambient
MDL Number: MFCD34859246
CAS Number: 1988359-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR