6-Bromo-3-chloro-2,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18576-1G 18576-500MG 18576-5G 18576-25G
77466-312EA 195.7 USD
77466-312 77479-056 77489-292 77499-656
6-Bromo-3-chloro-2,4-difluorobenzaldehyde ≥97%
6-Bromo-3-chloro-2,4-difluorobenzaldehyde

Precaución: For research use only

Formula: C₇H₂BrClF₂O
MW: 255.444 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845975
CAS Number: 1160573-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR