Ethyl-1-(cyanomethyl)-1H-pyrazole-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 1-(cyanomethyl)-1H-pyrazole-5-carboxylate

185554-1G 185554-500MG 185554-5G 185554-25G
77466-304EA 260.59 USD
77466-304 77479-048 77489-284 77499-652
Ethyl-1-(cyanomethyl)-1H-pyrazole-5-carboxylate ≥97%
Ethyl-1-(cyanomethyl)-1H-pyrazole-5-carboxylate

Precaución: For research use only

Formula: C₈H₉N₃O₂
MW: 179.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15146497
CAS Number: 1217862-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR