1-Bromo-4-ethoxy-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

18537-1G 18537-500MG 18537-5G 18537-25G
77466-300EA 318.27 USD
77466-300 77479-044 77489-280 77499-648
1-Bromo-4-ethoxy-2,5-dimethylbenzene ≥97%
1-Bromo-4-ethoxy-2,5-dimethylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21989013
CAS Number: 482355-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR