N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide ≥97%

Supplier: AOBChem USA

Warning

185015-1G 185015-500MG 185015-5G 185015-25G
77466-296EA 154.5 USD
77466-296 77479-040 77489-276 77499-644
N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide ≥97%
N-(6-Bromopyridin-2-yl)-2,2-dimethylpropanamide

Precaución: For research use only

Formula: C₁₀H₁₃BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD08235018
CAS Number: 221087-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR