2-Bromo-6-chloro-4-methylphenol ≥95%

Supplier: AOBChem USA

Warning

18447-1G 18447-500MG 18447-5G 18447-25G
77466-292EA 154.5 USD
77466-292 77479-036 77489-270 77499-634
2-Bromo-6-chloro-4-methylphenol ≥95%
2-Bromo-6-chloro-4-methylphenol

Precaución: For research use only

Formula: C₇H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD22414716
CAS Number: 57018-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR