tert-Butyl 3-(2-fluorophenyl)-3-hydroxypyrrolidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

184408-1G 184408-500MG 184408-5G
77466-288EA 798.25 USD
77466-288 77479-032 77489-266
tert-Butyl 3-(2-fluorophenyl)-3-hydroxypyrrolidine-1-carboxylate ≥95%
tert-Butyl 3-(2-fluorophenyl)-3-hydroxypyrrolidine-1-carboxylate

Precaución: For research use only

Formula: C₁₅H₂₀FNO₃
MW: 281.3226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28108557
CAS Number: 1421160-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR