2-(3,5-Difluorophenyl)-2-oxoacetic acid ≥97%

Supplier: AOBChem USA

Warning

184395-1G 184395-500MG 184395-5G 184395-25G
77466-284EA 186.43 USD
77466-284 77479-028 77489-262 77499-630
2-(3,5-Difluorophenyl)-2-oxoacetic acid ≥97%
2-(3,5-Difluorophenyl)-2-oxoacetic acid

Precaución: For research use only

Formula: C₈H₄F₂O₃
MW: 186.1123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07698681
CAS Number: 208259-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR