3-Bromo-4-methoxyphenyl acetate ≥95%

Supplier: AOBChem USA

Warning

18396-1G 18396-500MG 18396-5G 18396-25G
77466-276EA 95.79 USD
77466-276 77479-020 77489-256 77499-626
3-Bromo-4-methoxyphenyl acetate ≥95%
3-Bromo-4-methoxyphenyl acetate

Precaución: For research use only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529580
CAS Number: 124764-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR