1-(2,5-Dimethoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183623-1G 183623-500MG 183623-5G
77466-272EA 592.25 USD
77466-272 77479-016 77489-250
1-(2,5-Dimethoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-(2,5-Dimethoxyphenyl)-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₁H₉F₅O₃
MW: 284.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15145665
CAS Number: 1352207-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR