1-(2,5-Dimethylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183608-1G 183608-500MG 183608-5G
77466-268EA 592.25 USD
77466-268 77479-012 77489-246
1-(2,5-Dimethylphenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥95%
1-(2,5-Dimethylphenyl)-2,2,3,3,3-pentafluoro-1-propanone

Precaución: For research use only

Formula: C₁₁H₉F₅O
Storage Temperature: Ambient
MDL Number: MFCD15145691
CAS Number: 1352216-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR