2,2,3,3,3-Pentafluoro-1-[2-(1-methylethyl)phenyl]-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183579-1G 183579-500MG 183579-5G
77466-252EA 701.43 USD
77466-252 77478-996 77489-230
2,2,3,3,3-Pentafluoro-1-[2-(1-methylethyl)phenyl]-1-propanone ≥95%
2,2,3,3,3-Pentafluoro-1-[2-(1-methylethyl)phenyl]-1-propanone

Precaución: For research use only

Formula: C₁₂H₁₁F₅O
MW: 266.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15145671
CAS Number: 1443354-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR