2,2,3,3,3-Pentafluoro-1-(2-fluoro-5-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

183574-1G 183574-500MG 183574-5G 183574-25G
77466-248EA 415.09 USD
77466-248 77478-992 77489-226 77499-618
2,2,3,3,3-Pentafluoro-1-(2-fluoro-5-methylphenyl)-1-propanone ≥95%
2,2,3,3,3-Pentafluoro-1-(2-fluoro-5-methylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₀H₆F₆O
MW: 256.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34857752
CAS Number: 1930465-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR